missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bim Antibody (10B12) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-80103
This item is not returnable.
View return policy
Description
Bim Monoclonal specifically detects Bim in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Bim | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes an epitope within aa 20-40 of human, mouse, rat, monkey and dog BimL (∼19kDa), BimEL (∼23kDa), and BimS (∼16kDa). Also recognizes the six novel isoforms of human Bim, generated by alternative splicing and lacking the C-terminal hydrophobic region. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| 10B12 | |
| Unconjugated | |
| PBS containing 25% glycerol | |
| BCL2L11 | |
| Synthetic peptide corresponding to aa 20-40 of mouse BimL. | |
| 100 μg | |
| Apoptosis, Cancer, Immunology, Innate Immunity, Phospho Specific, Tumor Suppressors | |
| 10018 | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction