missing translation for 'onlineSavingsMsg'
Learn More

Bilobalide, MedChemExpress

Product Code. 16404747
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16404747 50 mg 50mg
16418976 100 mg 100mg
16408858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16404747 Supplier Medchem Express Supplier No. HYN0076
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bilobalide, a sesquiterpene trilactone constituent of Ginkgo biloba, inhibits the NMDA-induced efflux of choline with an IC50 value of 2.3 μM. Bilobalide prevents apoptosis through activation of the PI3K/Akt pathway in SH-SY5Y cells. Exerts protective and trophic effects on neurons.

Bilobalide, a sesquiterpene trilactone constituent of Ginkgo biloba, inhibits the NMDA-induced efflux of choline with an IC50 value of 2.3 μM. Bilobalide prevents apoptosis through activation of the PI3K/Akt pathway in SH-SY5Y cells. Exerts protective and trophic effects on neurons.

Specifications

Chemical Name or Material Bilobalide
CAS 33570-04-6
Color White
Purity Grade Notes Research
Molecular Formula C15H18O8
Quantity 50 mg
Synonym (-)-Bilobalide
Solubility Information DMSO : ≥ 100 mg/mL (306.47 mM)
SMILES O=C(O1)C[C@@]2([C@]1([H])C[C@]3(C(C)(C)C)O)[C@@]34[C@](OC([C@@H]4O)=O)([H])OC2=O
Molecular Weight (g/mol) 326.3
Formula Weight 326.3
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.