missing translation for 'onlineSavingsMsg'
Learn More

Biliverdine, TRC

Product Code. 17988144
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17988144 10 mg 10mg
17918614 50 mg 50mg
17939664 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17988144 Supplier Toronto Research Chemicals Supplier No. TRCB38640010MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114-25-0
Molecular Formula C33 H34 N4 O6
Molecular Weight (g/mol) 582.65
Synonym 21H-Biline-8,12-dipropanoic acid, 3,18-diethenyl-1,19,22,24-tetrahydro-2,7,13,17-tetramethyl-1,19-dioxo-, Biline-8,12-dipropionic acid, 1,19,22,24-tetrahydro-2,7,13,17-tetramethyl-1,19-dioxo-3,18-divinyl- (8CI), Pyrrole-3-propionic acid, 2-[[3-(2-carboxyethyl)-4-methyl-5-[(3-methyl-5-oxo-4-vinyl-3-pyrrolin-2-ylidene)methyl]-2H-pyrrol-2-ylidene]methyl]-4-methyl-5-[(4-methyl-5-oxo-3-vinyl-3-pyrrolin-2-ylidene)methyl]- (7CI), 3,18-Diethenyl-1,19,22,24-tetrahydro-2,7,13,17-tetramethyl-1,19-dioxo-21H-biline-8,12-dipropanoic acid, Biliverdin, Biliverdin IXα, Biliverdine, Dehydrobilirubin, NSC 62793, Oocyan, Protobiliverdin IXα, Uteroverdine, α-Biliverdin
IUPAC Name 3-[2-[(Z)-[3-(2-carboxyethyl)-5-[(Z)-(4-ethenyl-3-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methylpyrrol-2-ylidene]methyl]-5-[(Z)-(3-ethenyl-4-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-3-yl]propanoic acid
SMILES CC1=C(C=C)\C(=C\c2[nH]c(\C=C\3/N=C(\C=C\4/NC(=O)C(=C4C)C=C)C(=C3CCC(=O)O)C)c(CCC(=O)O)c2C)\NC1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C33H34N4O6/c1-7-20-19(6)32(42)37-27(20)14-25-18(5)23(10-12-31(40)41)29(35-25)15-28-22(9-11-30(38)39)17(4)24(34-28)13-26-16(3)21(8-2)33(43)36-26/h7-8,13-15,35H,1-2,9-12H2,3-6H3,(H,36,43)(H,37,42)(H,38,39)(H,40,41)/b26-13-,27-14-,28-15-
Formula Weight 582.2478
Chemical Name or Material Biliverdine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.