missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biliverdin Reductase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5111165
This item is not returnable.
View return policy
Description
Immunogen sequence: EPERKFGVVV VGVGRAGSVR MRDLRNPHPS SAFLNLIGFV SRRELGSIDG VQQISLEDAL SSQEVEVAYI CSESSSHEDY IRQFLNAGKH VL
Biliverdin reductase (BLVR) is a ubiquitously expressed, small cytoplasmic oxidoreductase that catalyzes the reduction of biliverdin to bilirubin. The first step of this process is catalyzed by heme oxygenase, an enzyme that converts heme to iron, carbon monoxide, and biliverdin. BLVR is unique among all enzymes to date in being dual pH/dual cofactor-dependent, as it requires NADH (pH 6.7) or NADPH (pH 8.7) as an electron donor. There are two isoforms of BLVR, biliverdin-IX alpha-reductase (BLVRA), the major component of human adult liver, and biliverdin-IX beta-reductase (BLVRB), found predominantly in fetal liver. The product of BLVR, bilirubin, is an important biological antioxidant, however it is also a neurotoxin and the cause of kernicterus. In addition to being an antioxidant, it inhibits kinase activity and the activity of enzymes such as protein kinase C and NADPH oxidase.
Specifications
| Biliverdin Reductase | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P53004 | |
| Blvra | |
| Recombinant Protein corresonding to Human Biliverdin Reductase | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Blvra | |
| 0610006A11Rik; 2500001N03Rik; Biliverdin reductase A; Biliverdin-IX alpha-reductase; BLVR; BLVRA; BVR; BVR A; BVRA; EC 1.3.1.24 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 644 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction