missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Biliverdin Reductase Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122198
This item is not returnable.
View return policy
Description
Biliverdin Reductase Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Biliverdin reductase (BLVR) is a ubiquitously expressed, small cytoplasmic oxidoreductase that catalyzes the reduction of biliverdin to bilirubin. The first step of this process is catalyzed by heme oxygenase, an enzyme that converts heme to iron, carbon monoxide, and biliverdin. BLVR is unique among all enzymes to date in being dual pH/dual cofactor-dependent, as it requires NADH (pH 6.7) or NADPH (pH 8.7) as an electron donor. There are two isoforms of BLVR, biliverdin-IX alpha-reductase (BLVRA), the major component of human adult liver, and biliverdin-IX beta-reductase (BLVRB), found predominantly in fetal liver. The product of BLVR, bilirubin, is an important biological antioxidant, however it is also a neurotoxin and the cause of kernicterus. In addition to being an antioxidant, it inhibits kinase activity and the activity of enzymes such as protein kinase C and NADPH oxidase.
Specifications
| Biliverdin Reductase | |
| Polyclonal | |
| Unconjugated | |
| Blvra | |
| 0610006A11Rik; 2500001N03Rik; Biliverdin reductase A; biliverdin reductase A-like protein; biliverdin-IX alpha-reductase; BLVR; Blvra; BVR; BVR A; BVRA; EC 1.3.1.24; testis tissue sperm-binding protein Li 61n | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 109778, 116599, 644 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P46844, P53004, Q9CY64 | |
| Blvra | |
| Recombinant protein Biliverdin Reductase A. The antigen corresponds to amino acid range 1-125 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction