missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biliverdin Reductase B/BLVRB Rabbit anti-Human, Clone: 9, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | Biliverdin Reductase B/BLVRB |
|---|---|
| Clone | 9 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18224158
|
Novus Biologicals
NBP2-90188-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18275147
|
Novus Biologicals
NBP2-90188-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Biliverdin Reductase B/BLVRB Monoclonal specifically detects Biliverdin Reductase B/BLVRB in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| Biliverdin Reductase B/BLVRB | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 645 | |
| IgG | |
| Protein A purified |
| 9 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Biliverdin reductase B, biliverdin reductase B (flavin reductase (NADPH)), Biliverdin-IX beta-reductase, BVR-B, EC 1.3.1.24, EC 1.5.1.30, flavin reductase, flavin reductase (NADPH), FLRBVRB, FR, GHBP, Green heme-binding protein, MGC117413, NADPH-dependent diaphorase, NADPH-flavin reductase, SDR43U1, short chain dehydrogenase/reductase family 43U, member 1 | |
| Recombinant Human Biliverdin Reductase B/BLVRB protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title