missing translation for 'onlineSavingsMsg'
Learn More

Biliverdin Reductase B/BLVRB Antibody [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30734209 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734209 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734209 Supplier Novus Biologicals Supplier No. AF6568J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Biliverdin Reductase B/BLVRB Polyclonal antibody specifically detects Biliverdin Reductase B/BLVRB in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Biliverdin Reductase B/BLVRB
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Biliverdin reductase B, biliverdin reductase B (flavin reductase (NADPH)), Biliverdin-IX beta-reductase, BVR-B, EC 1.3.1.24, EC 1.5.1.30, flavin reductase, flavin reductase (NADPH), FLRBVRB, FR, GHBP, Green heme-binding protein, MGC117413, NADPH-dependent diaphorase, NADPH-flavin reductase, SDR43U1, short chain dehydrogenase/reductase family 43U, member 1
Host Species Sheep
Immunogen E. coli-derived recombinant human Biliverdin Reductase B/BLVRB, Ala2-Gln206, predicted, Accession # P30043
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 645
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.