missing translation for 'onlineSavingsMsg'
Learn More

Biliverdin Reductase B/BLVRB Antibody, DyLight 350, Novus Biologicals™

Product Code. 18349526 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349526 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349526 Supplier Novus Biologicals Supplier No. NBP300081UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Biliverdin Reductase B/BLVRB Polyclonal specifically detects Biliverdin Reductase B/BLVRB in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Biliverdin Reductase B/BLVRB
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Biliverdin reductase B, biliverdin reductase B (flavin reductase (NADPH)), Biliverdin-IX beta-reductase, BVR-B, EC 1.3.1.24, EC 1.5.1.30, flavin reductase, flavin reductase (NADPH), FLRBVRB, FR, GHBP, Green heme-binding protein, MGC117413, NADPH-dependent diaphorase, NADPH-flavin reductase, SDR43U1, short chain dehydrogenase/reductase family 43U, member 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Biliverdin Reductase B/BLVRB (P30043; Ala2-Gln206).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 645
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.