missing translation for 'onlineSavingsMsg'
Learn More

Biliverdin Reductase B/BLVRB Antibody (2F4), Novus Biologicals™

Product Code. 18337567 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337567 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18337567 Supplier Novus Biologicals Supplier No. H00000645M09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Biliverdin Reductase B/BLVRB Monoclonal antibody specifically detects Biliverdin Reductase B/BLVRB in Human samples. It is validated for Western Blot, ELISA, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Biliverdin Reductase B/BLVRB
Applications Western Blot, ELISA, Functional Assay
Classification Monoclonal
Clone 2F4
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000704
Gene Alias Biliverdin reductase B, biliverdin reductase B (flavin reductase (NADPH)), Biliverdin-IX beta-reductase, BVR-B, EC 1.3.1.24, EC 1.5.1.30, flavin reductase, flavin reductase (NADPH), FLRBVRB, FR, GHBP, Green heme-binding protein, MGC117413, NADPH-dependent diaphorase, NADPH-flavin reductase, SDR43U1, short chain dehydrogenase/reductase family 43U, member 1
Host Species Mouse
Immunogen BLVRB (NP_000704, 107 a.a. ~ 206 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VACTSAFLLWDPTKVPPRLQAVTDDHIRMHKVLRESGLKYVAVMPPHIGDQPLTGAYTVTLDGRGPSRVISKHDLGHFMLRCLTTDEYDGHSTYPSHQYQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 645
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.