missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Biliverdin Reductase A/BLVRA Native Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
200 μg
50 μg
Unit Size:
100µg
200µg
50µg
Specifications
Specifications
| For Use With (Application) | SDS-PAGE, Western Blot |
| Formulation | 10mM Tris pH7.5, 0.1mM EDTA, 0.2mM DTT, 20% glycerol |
| Gene ID (Entrez) | 644 |
| Molecular Weight (g/mol) | M.W. Theoretical: 36 kDa |
| Quantity | 200 μg |
| Storage Requirements | Store at -70C. Avoid freeze-thaw cycles. |
| Gene Alias | biliverdin reductase A, Biliverdin-IX alpha-reductase, BLVR, BVR, BVR A, BVRA, EC 1.3.1.24 |
| Gene Symbol | BLVRA |
| Research Category | Cellular Signaling, metabolism, Protein Phosphatase, Serine Kinases |
| Purity or Quality Grade | >85% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction