missing translation for 'onlineSavingsMsg'
Learn More

Biliverdin Reductase A/BLVRA Antibody, Novus Biologicals™

Product Code. 18327281 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327281 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327281 Supplier Novus Biologicals Supplier No. NBP318247

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Biliverdin Reductase A/BLVRA Polyclonal antibody specifically detects Biliverdin Reductase A/BLVRA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Biliverdin Reductase A/BLVRA
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry 1:1000, Immunoprecipitation 1:100
Formulation PBS pH 7.4, 50% glycerol
Gene Alias biliverdin reductase A, Biliverdin-IX alpha-reductase, BLVR, BVR, BVR A, BVRA, EC 1.3.1.24
Host Species Rabbit
Immunogen Rat native full-length BVR purified from liver tissue
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Signaling, metabolism, Protein Phosphatase, Serine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 644
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.