missing translation for 'onlineSavingsMsg'
Learn More

Bilirubin Oxidase Antibody, Biotin, Novus Biologicals™

Product Code. 18324255 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324255 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324255 Supplier Novus Biologicals Supplier No. NBP300534

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Bilirubin Oxidase Polyclonal specifically detects Bilirubin Oxidase in Fungi samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bilirubin Oxidase
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot 1:500-1:2500, ELISA 1:1000-1:5000, Immunoprecipitation 1:100
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
Gene Alias Bilirubin oxidase, EC=1.3.3.5
Host Species Goat
Immunogen Bilirubin Oxidase [Myrothecium verrucaria] (Uniprot: Q12737)
Molecular Weight of Antigen 63.95 kDa
Purification Method Multi-step
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute with 100 ul deionized water (or equivalent)
Target Species Fungi
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.