missing translation for 'onlineSavingsMsg'
Learn More

Bilirubin, MedChemExpress

Product Code. 16434817
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434817 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16434817 Supplier Medchem Express Supplier No. HYN0323
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bilirubin is a yellow breakdown product of heme catabolism. Bilirubin exhibits antioxidant and antimutagenic effects.

Bilirubin is a yellow breakdown product of heme catabolism. Bilirubin exhibits antioxidant and antimutagenic effects.

Specifications

Chemical Name or Material Bilirubin
CAS 635-65-4
Color Orange
Purity Grade Notes Research
Molecular Formula C33H36N4O6
Quantity 50 mg
Solubility Information 0.1 M NaOH : 8.33 mg/mL (14.25 mM; ultrasonic and adjust pH to 10 with NaOH) ∣DMSO : 6 mg/mL (10.26 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.71 mM; ultrasonic and adjust pH to 11 with NaOH) ∣0.1 M HCL : < 1 mg/mL (insoluble)
SMILES CC(C(N/1)=O)=C(C=C)C1=C\C2=C(C)C(CCC(O)=O)=C(CC3=C(CCC(O)=O)C(C)=C(/C=C4NC(C(C=C)=C\4C)=O)N3)N2
Molecular Weight (g/mol) 584.66
Formula Weight 584.66
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.