missing translation for 'onlineSavingsMsg'
Learn More

Bilirubin 97%, Thermo Scientific Chemicals

Product Code. 11498137
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498137 1 g 1g
11408147 5 g 5g
11418147 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498137 Supplier Thermo Scientific Alfa Aesar Supplier No. A17522.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Bilirubin 97%, CAS # 635-65-4, is a tetrapyrroliccompound found in bile that is a product of heme catabolism.

Bilirubin is widely utilized to monitor the liver function and their disease, such as hepatitis or cirrhosis; or the effects of medicines which are damage the liver. It acts as a pigment in certain algae to capture light energy. Its double bonds are isomerizes in the presence of light, which is employed in phototherapy of jaundiced newborns babies. It is used to detect the blocking in bile ducts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Bilirubin is a degradation product of heme following the activity of heme oxidase. Bilirubin is produced in the reticuloendothelial system and conjugated in the liver. It appears as an orange-yellow pigment that is secreted in the bile
  • High levels of unconjugated bilirubin can result in jaundice, a condition of the yellowing of skin and eyes. This can be toxic and may be a sign of severe liver disease
  • Bilirubin has been seen to have functions including cell signaling, metabolism modulation, and immune regulation
  • It has protective antioxidant activity at moderate levels

Application

  • Bilirubin is widely used in studies of liver function and disease, such as hepatitis or cirrhosis
  • As an antioxidant, bilirubin has been studied in protection against H2O2-mediated cell death
  • It can be used as a scavenger of reactive oxygen species (ROS) or as an inhibitor of NADPH oxidase
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-65-4
Molecular Formula C33H36N4O6
Molecular Weight (g/mol) 584.67
MDL Number MFCD00005499
InChI Key BPYKTIZUTYGOLE-IFADSCNNSA-N
Synonym bilirubin, hematoidin, hemetoidin, bilirubin ix-alpha, principal bile pigment, unii-rfm9x3lj49, bilirubin ixalpha, 21h-biline-8,12-dipropanoic acid, 2,17-diethenyl-1,10,19,22,23,24-hexahydro-3,7,13,18-tetramethyl-1,19-dioxo, rfm9x3lj49, biline-8,12-dipropionic acid, 1,10,19,22,23,24-hexahydro-2,7,13,17-tetramethyl-1,19-dioxo-3,18-divinyl
PubChem CID 5280352
ChEBI CHEBI:16990
IUPAC Name 3-[2-[[3-(2-carboxyethyl)-5-[(Z)-(3-ethenyl-4-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-2-yl]methyl]-5-[(Z)-(4-ethenyl-3-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-3-yl]propanoic acid
SMILES CC1=C(C=C)\C(NC1=O)=C\C1=C(C)C(CCC(O)=O)=C(CC2=C(CCC(O)=O)C(C)=C(N2)\C=C2/NC(=O)C(C=C)=C2C)N1

Specifications

Quantity 1 g
Beilstein 74376
Sensitivity Light sensitive
Merck Index 14,1218
Solubility Information Soluble in chloroform,benzene,chlorobenzene,carbon disulfide,alkaline solutions and aqueous acid. Slightly soluble in ethanol and ether. Insoluble in water.
Formula Weight 584.68
Percent Purity 97%
Chemical Name or Material Bilirubin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.