missing translation for 'onlineSavingsMsg'
Learn More

Bilastine, MedChemExpress

Product Code. p-200100910
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16496466

Brand: Medchem Express HY14447

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bilastine is a selective histamine H1 receptor antagonist used for treatment of allergic rhinoconjunctivitis and urticaria.

Bilastine is a selective histamine H1 receptor antagonist used for treatment of allergic rhinoconjunctivitis and urticaria.

Specifications

Chemical Name or Material Bilastine
CAS 202189-78-4
Color White
Purity Grade Notes Research
Molecular Formula C28H37N3O3
Quantity 100 mg
Solubility Information DMSO : 10 mg/mL (21.57 mM; ultrasonic and warming and heat to 60°C)
SMILES CC(C)(C1=CC=C(CCN2CCC(C3=NC4=CC=CC=C4N3CCOCC)CC2)C=C1)C(O)=O
Molecular Weight (g/mol) 463.61
Formula Weight 463.61
Percent Purity 99.32%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.