missing translation for 'onlineSavingsMsg'
Learn More

Bilastine, MedChemExpress

Product Code. 16496466
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496466 100 mg 100mg
16416078 10 mg 10mg
16402217 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496466 Supplier Medchem Express Supplier No. HY14447

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bilastine is a selective histamine H1 receptor antagonist used for treatment of allergic rhinoconjunctivitis and urticaria.

Bilastine is a selective histamine H1 receptor antagonist used for treatment of allergic rhinoconjunctivitis and urticaria.

Specifications

Chemical Name or Material Bilastine
CAS 202189-78-4
Color White
Purity Grade Notes Research
Molecular Formula C28H37N3O3
Quantity 100 mg
Solubility Information DMSO : 10 mg/mL (21.57 mM; ultrasonic and warming and heat to 60°C)
SMILES CC(C)(C1=CC=C(CCN2CCC(C3=NC4=CC=CC=C4N3CCOCC)CC2)C=C1)C(O)=O
Molecular Weight (g/mol) 463.61
Formula Weight 463.61
Percent Purity 99.32%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.