missing translation for 'onlineSavingsMsg'
Learn More

Bikinin, MedChemExpress

Product Code. 16485906
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485906 100 mg 100mg
16471447 50 mg 50mg
16455238 10 mg 10mg
16447957 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485906 Supplier Medchem Express Supplier No. HY12524
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bikinin is a non-steroidal, ATP-competitive inhibitor of plant GSK-3/Shaggy-like kinases and activates BR (brassinosteroids) signaling.

Bikinin is a non-steroidal, ATP-competitive inhibitor of plant GSK-3/Shaggy-like kinases and activates BR (brassinosteroids) signaling.

Specifications

Chemical Name or Material Bikinin
CAS 188011-69-0
Color White
Purity Grade Notes Research
Molecular Formula C9H9BrN2O3
Quantity 100 mg
Synonym Abrasin
Solubility Information DMSO : ≥ 42 mg/mL (153.80 mM)
SMILES O=C(O)CCC(NC1=NC=C(Br)C=C1)=O
Molecular Weight (g/mol) 273.08
Formula Weight 273.08
Percent Purity 99.94%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.