missing translation for 'onlineSavingsMsg'
Learn More

BIIE-0246, MedChemExpress

Product Code. 16400969
Encompass_Preferred
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 16400969

Brand: Medchem Express HY101986

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress BIIE-0246 is a potent and highly selective non-peptide neuropeptide Y (NPY) Y2 receptor antagonist, with an IC50 of 15 nM.

BIIE-0246 is a potent and highly selective non-peptide neuropeptide Y (NPY) Y2 receptor antagonist, with an IC50 of 15 nM.

Specifications

Chemical Name or Material BIIE-0246
Physical Form Powder
CAS 246146-55-4
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C49H57N11O6
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym AR-H 053591
Solubility Information DMSO : 100 mg/mL (111.60 mM; Need ultrasonic)
SMILES O=C(N[C@H](C(NCCN1C(N(C2=CC=CC=C2)N(C3=CC=CC=C3)C1=O)=O)=O)CCCNC(N)=N)CC4(CC(N5CCN(C(C6=CC=CC=C67)C8=CC=CC=C8NC7=O)CC5)=O)CCCC4
Molecular Weight (g/mol) 896.05
Formula Weight 896.05
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.