missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Biglycan Recombinant Rabbit Monoclonal Antibody (23GB3700), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552626
This item is not returnable.
View return policy
Description
BGN is a small cellular or pericellular matrix proteoglycan that is closely related in structure to two other small proteoglycans, decorin and fibromodulin. The protein and decorin are thought to be the result of a gene duplication. Decorin contains one attached glycosaminoglycan chain, while this protein probably contains two chains. For this reason, this protein is called biglycan. This protein is thought to function in connective tissue metabolism by binding to collagen fibrils and transferring growth factor-beta. It may promote neuronal survival.
Specifications
| Biglycan | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P28653, P47853 | |
| BGN | |
| A synthetic peptide of human Biglycan (320-368AA). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3700 | |
| Unconjugated | |
| BGN | |
| BG; BGN; Biglycan; bone/cartilage proteclycan 1; bone/cartilage proteoglycan I; bone/cartilage proteoglycan-I; BSPG1; dermatan sulphate proteoglycan I; DSPG1; PG S1; PGI; PG-S1; SLRR1A; small leucine-rich protein 1A; Small proteoglycan I (biglycan), bone (BSPG1) (bone/cartilage proteclycan 1 precursor) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12111, 25181 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction