missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BigLEN (mouse)

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
BigLEN (mouse) is a GPR171 agonist. ProSAAS-derived neuropeptide. Regulates food intake in mice. Inhibits the release of glutamate onto parvocellular neurons of the paraventricular nucleus in a process dependent upon activation of postsynaptic G proteins.
Specifications
Specifications
| CAS | 501036-69-7 |
| Content And Storage | Store at -20°C |
| Molecular Weight (g/mol) | 1756.03 |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction