missing translation for 'onlineSavingsMsg'
Learn More

Bigfoot™ Spectral Sorting and Analysis Package for Sasquatch Software

Product Code. 18861362
Change view
Click to view available options
For Use With (Equipment):
Bigfoot Spectral Cell Sorter
Product Code. For Use With (Equipment)
18861362 Bigfoot Spectral Cell Sorter
1 options
This item is not returnable. View return policy

Product Code. 18861362

Brand: THERMO SCIE LSG CUSTOM PL00322

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Invitrogen Bigfoot Spectral Cell Sorter provides power, performance, and flexibility to accelerate cutting edge discoveries.

The addition of this Spectral Sorting and Analysis Package for Sasquatch Software (SQS) allows the operator to sort and analyze either spectrally unmixed data or traditional compensated data on the Bigfoot cell sorter. This option must be added to the instrument to enable spectral sorting and analysis.

This Spectral Sorting and Analysis Package provides full spectrum data using up to 55 detectors for 30+ colors.Including the spectral capabilities on the Bigfoot sorter enables:

  • Experimental design tools with complexity indices and a spectral similarity matrix
  • Automated protocol and spectral plot generation for running controls
  • Subtraction of auto-fluorescence signal from experimental samples
  • Real-time spectral unmixing of samples during acquisition or sorting
TRUSTED_SUSTAINABILITY

Specifications

Format Software Application
For Use With (Equipment) Bigfoot Spectral Cell Sorter

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.