Learn More
Invitrogen™ Bigfoot™ Spectral Cell Sorter Accessories
Description
The Invitrogen Bigfoot Spectral Cell Sorter provides power, performance, and flexibility to help accelerate cutting edge discoveries. Designed to streamline the cell sorting workflow and enhance the user experience, the Bigfoot sorter has a multitude of cell sorting advancements, accessories, and unique capabilities to enable the most complex multi-parameter sorts and to meet high-throughput demands for a wide range of applications.
The Bigfoot Spectral Cell Sorter offers a full line of accessory products for continuous expansion of the instrument, including interchangeable nozzle tips, temperature controlled sample input and sort output adapters, and bulk fluid bottles and caps.
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| For Use With (Equipment) | Bigfoot™ Spectral Cell Sorter |
| For Use With (Application) | Flow Cytometry |
| Quantity | Each |
| Content And Storage | Room Temperature |
| Product Type | Output Tube Adapter, 50 mL |
| Product Line | Bigfoot |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.