missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ BigDye XTerminator™ Purification Kit

Product Code. 15741826 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 Preps
1000 Preps
2500 Preps
40,000 Preps
Unit Size:
1000 reactions
2mL
Each
Pack of 100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15741826 100 Preps 2mL
15929202 100 Preps Pack of 100
15899816 1000 Preps Each
16315312 1000 Preps 1000 reactions
15879816 2500 Preps Each
15889816 40,000 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15741826 Supplier Applied Biosystems™ Supplier No. 4376486
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

2 mL XTerminator™ Solution (store at 4°C), 9 mL SAM™ Solution (store at room temperature or 4°C)

The BigDye XTerminator™ Purification Kit is a fast, simple purification method for DNA sequencing reactions that removes unincorporated BigDye™ terminators and salts.

  • Eliminate liquid transfer - Use a single plate
  • Obtain complete dye blob removal
  • Stabilize the samples before analysis
  • Improve your sequencing workflow
  • Perform manual or automated protocols
  • Get rapid, reliable, and reproducible results

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 mL XTerminator™ Solution (store at 4°C), 9 mL SAM™ Solution (store at room temperature or 4°C)
Format Kit
Sample Type Sequencing Products
Final Product Type DNA
For Use With (Application) Sequencing
Includes 2 Bottles
No. of Reactions 100
Product Line BigDye XTerminator, BigDye
Product Type Purification Kit
Quantity 100 Preps
Shipping Condition Wet Ice
Target Sequencing Reactions
Test Time 40 min
Starting Material Amount 10 μl
Show More Show Less
compliance-icons
Product Identifier
  • BigDye XTerminator Purification Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: benzene, dowex MR-3mixed ion exchange resin

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.