missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ 3730/3730xl DNA Analyzer Sequencing Standards, BigDye™ Terminator v3.1
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
Includes
Kit contains 4 tubes of hsp69 standard sequence used for instrument and spectral calibrations
This kit contains four tubes of hsp69 standard sequence used for instrument and spectral calibrations.
Order Info
Kit
Specifications
Specifications
| Content And Storage | This kit contains 4 tubes of hsp69 standard sequence used for instrument and spectral calibrations. This material is used with the 3730, and 3730xl DNA Analyzers. Store kit at -15 to -25°C. |
| Format | Tube |
| For Use With (Equipment) | 3730xl DNA Analyzer, 3730 DNA Analyzer |
| For Use With (Application) | Sequencing |
| Product Line | BigDye Terminator, BigDye |
| Product Type | Sequencing Standards |
| Quantity | 1 Kit |
| Shipping Condition | Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction