missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 3730/3730xl DNA Analyzer Sequencing Standards, BigDye™ Terminator v3.1

Product Code. 15819786 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15819786 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819786 Supplier Applied Biosystems™ Supplier No. 4336943
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Kit contains 4 tubes of hsp69 standard sequence used for instrument and spectral calibrations

This material is used with the 3730, and 3730xl DNA Analyzers.

This kit contains four tubes of hsp69 standard sequence used for instrument and spectral calibrations.

Order Info

Kit

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This kit contains 4 tubes of hsp69 standard sequence used for instrument and spectral calibrations. This material is used with the 3730, and 3730xl DNA Analyzers. Store kit at -15 to -25°C.
Format Tube
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer
For Use With (Application) Sequencing
Product Line BigDye Terminator, BigDye
Product Type Sequencing Standards
Quantity 1 Kit
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.