Learn More
Applied Biosystems™ BigDye™ Terminator v3.1 Cycle Sequencing Kit
The BigDye™ Terminator v3.1 Cycle Sequencing Kit's robust, highly flexible chemistry
Brand:  Applied Biosystems™ 4337458
Additional Details : Weight : 0.00450kg
 View more versions of this product
Product Code. 15819796
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes: 10 x 20mL Tubes of BigDye™ Terminator v3.1 Ready Reaction Mix, 4 tubes M13 (-21) Primer, 4 tubes pGEM Control DNA, 1 x 233mL tube of 5X Sequencing Buffer (may be stored at 4°C)
Description
Description
  • Improve the quality of your results for a wide range of sequencing applications
  • Optimized for long read lengths
  • Better dye mobility characteristics
  • Improved performance reading through GT rich regions
  • Get longer, higher-quality reads with more uniform peak heights and optimal signal balance
  • Enhance your productivity and reduce costs
Order Info
Each
Specifications
Specifications
• 10 × 20-ml tubes of BigDye™ Terminator v3.1 Ready Reaction Mix
• 4 tubes M13 (-21) Primer
• 4 tubes pGEM Control DNA
• 1 × 233-ml tube of 5X Sequencing Buffer (may be stored at 4°C)
Store kit at -15°C to -25°C, and avoid repeated freeze-thaw cycles.
12-well Strips, 8-well Strip, 96- or 384-well Plate
Standard Sequencing
25,000
Sequencing Buffer
BAC DNA, Plasmid DNA (≤15Kb), PCR Amplicons, Single Stranded DNA
Cycle Sequencing Kit, 25,000 rxns, Includes 10 x 20mL tubes of BigDye™ Terminator v3.1 Ready Reaction Mix, 4 tubes M13 (-21) Primer, 4 tubes pGEM Control DNA, 1 x 233mL tube of 5X Sequencing Buffer (may be stored at 4°C)
310 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, GeneAmp 9700, Veriti Thermal Cycler, 3130 Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
10 x 20mL tubes of BigDye™ Terminator v3.1 Ready Reaction Mix, 4 tubes M13 (-21) Primer, 4 tubes pGEM Control DNA, 1 x 233mL tube of 5X Sequencing Buffer (may be stored at 4°C)
BigDye™ Terminator, BigDye™
Fluorescent Dye Terminator Sequencing