missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Matrix Standards Kit, BigDye™ Terminator v3.1, for 31xx/3500/SeqStudio™/SeqStudio™ Flex

Product Code. 15906065 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15906065

Brand: Applied Biosystems™ 4336974

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

This kit contains two tubes of standard used to generate a spectrum required when analyzing BigDye Terminator v3.1 labeled DNA fragments on 31xx, 3500, SeqStudio, and SeqStudio Flex series genetic analyzers. Each tube contains a blend of four different colors of fluorescent dye-labeled DNA fragments suspended in 1X TE buffer.

This kit contains two tubes of standard used to generate a spectrum required when analyzing BigDye Terminator v3.1 labeled DNA fragments on 31xx, 3500, SeqStudio, and SeqStudio Flex series genetic analyzers.

This kit contains two tubes of standard used to generate a spectrum required when analyzing BigDye Terminator v3.1 labeled DNA fragments on 31xx, 3500, SeqStudio, and SeqStudio Flex series genetic analyzers. Each tube contains a blend of four different colors of fluorescent dye-labeled DNA fragments suspended in 1X TE buffer.

Order Info

1 Kit

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2–8°C protected from light. Do not freeze.
Format Kit
For Use With (Equipment) 31xx Series Genetic Analyzers, SeqStudio™ Genetic Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
For Use With (Application) Sequencing
Label or Dye ROX, TAMRA™, Rhodamine 110, Rhodamine 6G
Product Line BigDye™ Terminator, BigDye™
Product Type Matrix Standard
Quantity 1 kit
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.