missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ 310 Genetic Analyzer Matrix Standards, BigDye™ Terminator v1.1
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
Includes
Each kit contains a set of four standards used to generate a matrix file for the 310 Genetic Analyzer. Each tube contains a different fluorescent dye in solution with blue loading dye. Contents are stable for 6 months at 2 to 8°C. Do not freeze
Each tube contains a different fluorescent dye in solution, along with blue loading dye.
Order Info
Each
Specifications
Specifications
| Content And Storage | Each kit contains a set of four standards used to generate a matrix file for the 310 Genetic Analyzer. Each tube contains a different fluorescent dye in solution with blue loading dye. Contents are stable for 6 months at 2 to 8°C. Do not freeze. |
| For Use With (Equipment) | 310 Genetic Analyzer |
| For Use With (Application) | Sequencing |
| Label or Dye | ROX, TAMRA™, Rhodamine 110, Rhodamine 6G |
| Product Line | BigDye™ Terminator, BigDye™ |
| Product Type | Sequencing Standards |
| Quantity | 1 Kit |
| Shipping Condition | Wet Ice |
For research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction