missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Matrix Standards Kit, BigDye™ Terminator v1.1, for 31xx/3500/SeqStudio™/SeqStudio™ Flex

Product Code. 15996055 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Product Code. Quantity unitSize
15996055 1 kit 1 set
1 options
This item is not returnable. View return policy

Product Code. 15996055

Brand: Applied Biosystems™ 4336824

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This kit contains two tubes of standard used to generate a spectrum required when analyzing BigDye Terminator v1.1 labeled DNA fragments on the 31xx, 3500, SeqStudio™, and SeqStudio Flex series genetic analyzers.

This kit contains two tubes of standard used to generate a spectrum required when analyzing BigDye Terminator v1.1 labeled DNA fragments on the 31xx, 3500, SeqStudio, and SeqStudio Flex series genetic analyzers. Each tube contains a blend of four different colors of fluorescent dye-labeled DNA fragments suspended in 1X TE buffer.

Order Info

1 Kit

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2–8°C protected from light. Do not freeze.
Format Kit
For Use With (Equipment) 31xx Series Genetic Analyzer, SeqStudio™ Genetic Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
For Use With (Application) Sequencing
Label or Dye ROX, TAMRA™, Rhodamine 110, Rhodamine 6G
Product Line BigDye Terminator, BigDye
Product Type Matrix Standard
Quantity 1 kit
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.