missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Matrix Standards Kit, BigDye™ Terminator v1.1, for 31xx/3500/SeqStudio™/SeqStudio™ Flex

Product Code. 15711826 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15711826 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15711826 Supplier Applied Biosystems™ Supplier No. 4336824

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This kit contains two tubes of standard used to generate a spectrum required when analyzing BigDye Terminator v1.1 labeled DNA fragments on the 31xx, 3500, SeqStudio™, and SeqStudio Flex series genetic analyzers.

This kit contains two tubes of standard used to generate a spectrum required when analyzing BigDye Terminator v1.1 labeled DNA fragments on the 31xx, 3500, SeqStudio, and SeqStudio Flex series genetic analyzers. Each tube contains a blend of four different colors of fluorescent dye-labeled DNA fragments suspended in 1X TE buffer.

Beställningsinfo

1 Kit

TRUSTED_SUSTAINABILITY

Specifikationer

Content And Storage Store at 2–8°C protected from light. Do not freeze.
Format Kit
For Use With (Equipment) 31xx Series Genetic Analyzer, SeqStudio™ Genetic Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
For Use With (Application) Sequencing
Label or Dye ROX, TAMRA™, Rhodamine 110, Rhodamine 6G
Product Line BigDye Terminator, BigDye
Product Type Matrix Standard
Quantity 1 kit
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.