missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ BDT/POP/Buffer Bundle

Product Code. 30534944 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Reactions:
96 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
30534944 96 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30534944 Supplier Applied Biosystems™ Supplier No. A66292
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

The BDT/POP/Buffer Bundle includes a BigDye Terminator 3.1 Cycle Sequencing Kit, POP-7 Polymer, Anode Buffer Containers, and Cathode Buffer Containers for your 3500 or SeqStudio Flex genetic analyzer. All products are offered in a single, easy-to-order bundle to provide key reagents for Sanger sequencing workflows.

The BDT/POP/Buffer Bundle includes a BigDye Terminator 3.1 Cycle Sequencing Kit, POP-7 Polymer, Anode Buffer Containers, and Cathode Buffer Containers for your 3500 or SeqStudio Flex genetic analyzer. All products are offered in a single, easy-to-order bundle to provide key reagents for Sanger sequencing workflows.

BigDye Terminator v3.1 Cycle Sequencing Kit

The BigDye Terminator v3.1 Cycle Sequencing Kit provides premixed reagents for Sanger sequencing reactions. The kit's robust, highly flexible chemistry is ideal for de novo sequencing, resequencing, and finishing with PCR product, plasmid, fosmid, and BAC templates.

  • Improve the quality of your results for a wide range of sequencing applications
  • Optimized for long read lengths
  • Better dye mobility characteristics
  • Improved performance reading through GT-rich regions
  • Get longer, higher-quality reads with more uniform peak heights and optimal signal balance
  • Enhance your productivity and reduce costs

POP-7 Polymer

POP-7 Polymer is a separation matrix for the 3500 and SeqStudio Flex DNA analyzers. It is customized for DNA sequencing and fragment analysis.

  • Easy to use—pre-formulated and in a single-use pouch, which not only saves time but helps ensure reliability and consistency
  • Includes radio frequency identification (RFID) label—easy to track important information such as part⁄lot number, samples remaining, and expiry dates (displayed on software dashboard)
  • Robust formulation—formulated for sequencing and fragment analysis applications
  • Replenishable—allow capillaries to be used multiple times

Cathode Buffer Containers

The Cathode Buffer Container (CBC) contains 1X running buffer to support all electrophoresis applications on 3500 and SeqStudio Flex series genetic analyzers. The container has two separate compartments: the left compartment provides the cathode buffer for electrophoresis and the right provides for a capillary wash and spent polymer waste ejection functionality between injections. The CBC is made in a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the CBC is heat sealed with a plastic film, which should be removed prior to direct installation on to the instrument. Each package includes four individual containers.

Anode Buffer Containers

The Anode Buffer Container (ABC) contains 1X running buffer to support all electrophoresis applications on the 3500 and SeqStudio Flex series genetic analyzers. It is made in a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the ABC is heat sealed with a plastic film, which should be removed prior to direct installation on the instrument. Each package includes four individual containers.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • BigDye Terminator v3.1 Ready Reaction Mix, 1 x 800 μL (Store at -15°C to -25°C)
• M13 (-21) Primer, 1 tube (-15°C to -25°C)
• pGEM Control DNA, 1 tube (-15°C to -25°C)
• 5X Sequencing Buffer, 1 x 12 mL (-15°C to -25°C, or 2°C to 8°C)
• POP-7 Polymer, 96 reactions (2°C to 8°C)
• Cathode buffer container, 4 packs (2°C to 8°C)
• Anode buffer container, 4 packs (2°C to 8°C)
Control Type Plasmid DNA
Detection Method Fluorescent Dye
Format Kit
Isolation Technology Enzymatic, Magnetic Bead
Sample Type Purified DNA
Sequencing Type Genome and DNA Sequencing
Enzyme Proprietary
For Use With (Equipment) 3500xL Genetic Analyzer, 3500 Genetic Analyzer
For Use With (Application) Standard Sequencing
Label or Dye Various
No. of Reactions 96 Reactions
Product Type Sequencing Buffer
Quantity 96 Reactions
Species Various
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.