missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ BigDye™ Direct Cycle Sequencing Kit
Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
100 Reactions
1000 Reactions
24 Reactions
Unit Size:
100 reactions
1000 reactions
24 reactions
Description
Includes
This kit contains BigDye™ Direct PCR Master Mix, Sequencing Master Mix, M13 Fwd Primer, M13 Rev Primer, and Control DNA CEPH 1347-02, This kit contains sufficient reagents for 24 reactions
- A streamlined, simple procedure
- Performance of all sequencing steps in a single reaction tube
- Fast migration with excellent resolution starting at the 1st base after the gene-specific primer sequence
- Substantial time savings versus previous workflows
Order Info
1 Kit
Specifications
Specifications
| Content And Storage | This kit contains BigDye™ Direct PCR Master Mix, Sequencing Master Mix, M13 Fwd Primer, M13 Rev Primer, and Control DNA CEPH 1347-02. Store all components at -15 to -25°C and protect from light. PCR Master Mix should be stored at 4°C after first use. This kit contains sufficient reagents for 24 reactions. |
| Format | 8-well Strip, 96-well Plate |
| For Use With (Equipment) | 3130xl Genetic Analyzer, 3730xl DNA Analyzer, GeneAmp 9700, SeqStudio Genetic Analyzer, Veriti Thermal Cycler, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer |
| For Use With (Application) | Comparative Sequencing (Germline Mutations - 30:70 Heterozygotes), Comparative Sequencing (Germline Mutations - 50:50), GC-Rich Sequencing (>65%), GT-Rich or Difficult Template Sequencing, Standard Sequencing, Gap Closure, Homopolymer A or T, AT-Rich Sequencing (>65%), Comparative Sequencing (Germline Mutations -10:90 Heterozygotes) |
| Includes | BigDye™ Direct Cycle Sequencing Kit, 24 rxns, 8-well Strips, 96-well Plate, Fluorescent Dye Terminator Sequencing Technique, 3, Use with 130xl Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, GeneAmp™ 9700, Veriti™ Thermal Cycler |
| No. of Reactions | 24 Reactions |
| Product Line | BigDye |
| Product Type | Direct Cycle Sequencing Kit |
| Quantity | 24 reactions |
| Shipping Condition | Dry Ice |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction