Learn More
Trajan Scientific and Medical Big Trap - universal
Product Code. p-7158664
Click to view available options
Port Type:
1/4 in.
1/8 in.
Unit Size:
Each
Product Code. 15760979
Brand: Trajan Scientific and Medical undefined
Only null left
Add to basket
Add to basket
For bulk purification applications or where several instruments are plumbed from a single source, a Big Trap gas filter is an ideal solution. A one-piece heavy walled alminium tube provides 750cm3 of capacity and a pressure rating of up to 250 psig
  • High capacity filters ideal for bulk purification - cumulative capacity level of 100μg/L of oxygen, moisture, hydrocarbons, carbon dioxide and carbon monoxide at a flow rate of 8L/min
  • Efficiency < 100 ppb
  • Can be used where several instruments are plumbed from a single gas source
  • Sintered stainless steel frits prevent particulate contamination
Specifications
For Use With (Application) GC and GCMS applications where helium is employed
Model Hydrocarbon
Volume (Metric) 750 cm3
Port Type 1/8 in.
Description Removes oxygen, moisture, hydrocarbons, carbon dioxide and carbon monoxide
For Use With Big Trap Mounting Clip
Pressure 250 psig