missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Big ET-1 ELISA Kit
Description
The Human Big ET-1 Platinum ELISA is an enzyme-linked immunosorbent assay for the quantitative detection of human Big ET-1. Cell culture supernatant, serum, and plasma (EDTA, citrate, heparin) have been tested with this assay.
ELISA Kit
Specifications
Specifications
| Detection Method | Colorimetric |
| Product Type | ELISA Kit |
| Target Species | Human |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Quantity | 960 Tests |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?