missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Bifonazole, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Bifonazole |
| CAS | 60628-96-8 |
| Molecular Formula | C22 H18 N2 |
| Quantity | 1 g |
| Synonym | 1-([1,1'-Biphenyl]-4-ylphenylmethyl)-1H-imidazole, (±)-Bifonazole, A-One-L, Amycor, Azolmen, BAY-h 4502, Bedriol, Bicutrin, Bifazol, Bifonazole, Mycospor, Mycosporan, Trifonazole, bifosin |
| InChI Formula | InChI=1S/C22H18N2/c1-3-7-18(8-4-1)19-11-13-21(14-12-19)22(24-16-15-23-17-24)20-9-5-2-6-10-20/h1-17,22H |
| SMILES | c1ccc(cc1)C(c2ccc(cc2)c3ccccc3)n4ccnc4 |
| IUPAC Name | 1-[phenyl-(4-phenylphenyl)methyl]imidazole |
| Molecular Weight (g/mol) | 310.39 |
| Formula Weight | 310.147 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction