missing translation for 'onlineSavingsMsg'
Learn More

Bifonazole, Mikromol™

Product Code. 30052173
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 30052173

Brand: Mikromol MM0318.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bifonazole
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bifonazole
CAS 60628-96-8
Analyte or Component Names Bifonazole
Shipping Condition Room Temperature
Molecular Formula C22 H18 N2
Certifications/Compliance ISO 17034
Synonym 1-([1,1'-Biphenyl]-4-ylphenylmethyl)-1H-imidazole, A-One-L, Amycor, Azolmen, BAY-h 4502, Bedriol, Bicutrin, Bifazol, Bifonazole, Mycospor, Mycosporan, Trifonazole, bifosin, (+/-)-Bifonazole
InChI Formula InChI=1S/C22H18N2/c1-3-7-18(8-4-1)19-11-13-21(14-12-19)22(24-16-15-23-17-24)20-9-5-2-6-10-20/h1-17,22H
SMILES c1ccc(cc1)C(c2ccc(cc2)c3ccccc3)n4ccnc4
IUPAC Name 1-[phenyl-(4-phenylphenyl)methyl]imidazole
Molecular Weight (g/mol) 310.39
Quantity 500 mg
Formula Weight 310.147 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.