Learn More
Bifonazole, 99%, Thermo Scientific™
£40.40 - £125.00
Chemical Identifiers
| CAS | 60628-96-8 |
|---|---|
| Molecular Formula | C22H18N2 |
| Molecular Weight (g/mol) | 310.4 |
| MDL Number | MFCD00865567 |
| InChI Key | OCAPBUJLXMYKEJ-UHFFFAOYSA-N |
| Synonym | Mycosporan; 1-(alpha,4-Diphenylbenzyl)imidazole |
| PubChem CID | 2378 |
| ChEBI | CHEBI:78692 |
| IUPAC Name | 1-[phenyl-(4-phenylphenyl)methyl]imidazole |
| SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15485519
|
Thermo Scientific Alfa Aesar
J63253.03 |
1g |
£40.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15495519
|
Thermo Scientific Alfa Aesar
J63253.06 |
5g |
£125.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsBifonazole is an imidazole antifungal agent and calmodulin antagonist. It causes a reduction in glycolysis and ATP levels in B16 melanoma cells. Bifonazole is an inhibitor of C14orf1. Bifonazole is an imidazole-based anti-fungal agent with broad spectrum activity against many fungi, molds, yeast and some gram-positive bacteria. Bifonazole inhibits ergosterol biosynthetic protein 28 and Cytochrome P450 2B4.
Solubility
Soluble in DMSO
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 60628-96-8 | |
| 310.4 | |
| OCAPBUJLXMYKEJ-UHFFFAOYSA-N | |
| 2378 | |
| 1-[phenyl-(4-phenylphenyl)methyl]imidazole |
| C22H18N2 | |
| MFCD00865567 | |
| Mycosporan; 1-(alpha,4-Diphenylbenzyl)imidazole | |
| CHEBI:78692 | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4 |
Specifications
| 60628-96-8 | |
| C22H18N2 | |
| 14,1213 | |
| Soluble in DMSO | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4 | |
| 310.4 | |
| CHEBI:78692 | |
| 99% |
| White | |
| MFCD00865567 | |
| Mycosporan; 1-(alpha,4-Diphenylbenzyl)imidazole | |
| OCAPBUJLXMYKEJ-UHFFFAOYSA-N | |
| 1-[phenyl-(4-phenylphenyl)methyl]imidazole | |
| 2378 | |
| 310.39 | |
| Bifonazole |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 262-336-6
missing translation for 'rtecsNumber' : NI3517000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.