missing translation for 'onlineSavingsMsg'
Learn More

Bifenox, TRC

Product Code. 30345605
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30345605 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345605 Supplier Toronto Research Chemicals Supplier No. TRCB38290050MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bifenox
CAS 42576-02-3
Molecular Formula C14 H9 Cl2 N O5
Quantity 50 mg
Synonym 2,4-Dichlorophenyl 3-(methoxycarbonyl)-4-nitrophenyl ether, 2,4-Dichlorophenyl 3'-carbomethoxy-4'-nitrophenyl ether, 2,4-Dichlorophenyl-3'-methoxycarbonyl-4'-nitrophenyl ether, 3-Carbomethoxy-2',4'-dichloro-4-nitrodiphenyl ether, Bifenox, Biphenox, Fox, Jiazhichucaomi, MC 4379, MC 79, Methyl 5-(2,4-dichlorophenoxy)-2-nitrobenzoate, Modown, Modown 4F, Weeral
InChI Formula InChI=1S/C14H9Cl2NO5/c1-21-14(18)10-7-9(3-4-12(10)17(19)20)22-13-5-2-8(15)6-11(13)16/h2-7H,1H3
SMILES COC(=O)c1cc(Oc2ccc(Cl)cc2Cl)ccc1[N+](=O)[O-]
IUPAC Name methyl 5-(2,4-dichlorophenoxy)-2-nitrobenzoate
Molecular Weight (g/mol) 342.13
Formula Weight 340.9858
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.