missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bif1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553340
This item is not returnable.
View return policy
Description
Immunogen sequence: DRKAPSRINN PELLGQYMID AGTEFGPGTA YGNALIKCGE TQKRIGTADR ELIQTSALNF LTPLRNFIEG DYKTIAKER Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Bif1 gene encodes a protein similar to a protein in rodents which is induced by bone morphogenic protein 2 in vitro and may be involved in BMP2-induced transcription.
Specifications
| Bif1 | |
| Polyclonal | |
| Unconjugated | |
| Sh3glb1 | |
| AA409932; AI314629; AU015566; Bax-interacting factor 1; BIF1; Bif-1; CGI-61; dJ612B15.2; EMBL:AAH79085.1}; endophilin b1; endophilin-B1; Kiaa0491; mKIAA0491; PPP1R70; protein phosphatase 1, regulatory subunit 70; SH3 domain containing GRB2 like endophilin B1; SH3 domain-containing GRB2-like protein B1; SH3-containing protein SH3GLB1; SH3-domain GRB2 like endophilin B1; SH3-domain GRB2-like B1 (endophilin); SH3-domain GRB2-like endophilin B1; SH3GLB1; sh3glb1 {ECO:0000312; testicular tissue protein Li 172 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51100 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y371 | |
| Sh3glb1 | |
| Recombinant protein corresponding to Human Bif1. Recombinant protein control fragment (Product #RP-90706). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction