missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bif1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12818412
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12818412 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12818412 Supplier Invitrogen™ Supplier No. PA520248

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Hela cell lysate. PA5-20248 can be used with blocking peptide PEP-0370.

Bif1 gene encodes a protein similar to a protein in rodents which is induced by bone morphogenic protein 2 in vitro and may be involved in BMP2-induced transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bif1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Sh3glb1
Gene Accession No. Q9JK48, Q9Y371
Gene Alias AA409932; AI314629; AU015566; Bax-interacting factor 1; BIF1; Bif-1; CGI-61; dJ612B15.2; EMBL:AAH79085.1}; endophilin b1; endophilin-B1; Kiaa0491; mKIAA0491; PPP1R70; protein phosphatase 1, regulatory subunit 70; SH3 domain containing GRB2 like endophilin B1; SH3 domain-containing GRB2-like protein B1; SH3-containing protein SH3GLB1; SH3-domain GRB2 like endophilin B1; SH3-domain GRB2-like B1 (endophilin); SH3-domain GRB2-like endophilin B1; SH3GLB1; sh3glb1 {ECO:0000312; testicular tissue protein Li 172
Gene Symbols Sh3glb1
Host Species Rabbit
Immunogen A 15 amino acid peptide from near the carboxy terminus of human BIF.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51100, 54673
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.