missing translation for 'onlineSavingsMsg'
Learn More

Biebrich Scarlet sodium salt

Product Code. 15496989
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15496989

Brand: Thermo Scientific Alfa Aesar J65603.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4196-99-0 | C22H14N4Na2O7S2 | 556.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4196-99-0
Molecular Formula C22H14N4Na2O7S2
Molecular Weight (g/mol) 556.47
MDL Number MFCD00003891
InChI Key YCKMEQRBEVVZQF-VKZXTWAVSA-L
Synonym C.I. 26905; Acid Red 66
PubChem CID 16219040
IUPAC Name disodium 2-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-[2-(4-sulfonatophenyl)diazen-1-yl]benzene-1-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(=C1)S([O-])(=O)=O

Specifications

Melting Point 181°C to 188°C
Beilstein 3883900
Formula Weight 556.49
Color Brown to Red
Quantity 25 g
Flash Point 300°C (572°F)
Physical Form Powder
Chemical Name or Material Biebrich Scarlet sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.