missing translation for 'onlineSavingsMsg'
Learn More

BID Antibody (002), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18417785 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417785 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417785 Supplier Novus Biologicals Supplier No. NBP289515JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BID Monoclonal antibody specifically detects BID in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen BID
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 002
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID, tbid
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human BID (Uniprot#: P55957-1; Met1-Asp195).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 637
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.