missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BID Antibody (002), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89515H
This item is not returnable.
View return policy
Description
BID Monoclonal specifically detects BID in Human samples. It is validated for Western Blot, ELISA.
Specifications
| BID | |
| Monoclonal | |
| HRP | |
| apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID, tbid | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human BID (P55957-1; Met1-Asp195). | |
| 0.1 mL | |
| Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
| 637 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 2 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Has cross-reactivity in ELISA with: Mouse BID | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction