missing translation for 'onlineSavingsMsg'
Learn More

Bicyclol, MedChemExpress

Product Code. 16463796
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463796 25 mg 25mg
16434477 50 mg 50mg
16431058 5 mg 5mg
16408608 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16463796 Supplier Medchem Express Supplier No. HYB0766
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bicyclol(SY 801) is a anti-hepatitis drug.

Bicyclol(SY 801) is a anti-hepatitis drug.

Specifications

Chemical Name or Material Bicyclol
CAS 118159-48-1
Color White
Purity Grade Notes Research
Molecular Formula C19H18O9
Quantity 25 mg
Synonym SY801
Solubility Information DMSO : 100 mg/mL (256.19 mM; Need ultrasonic)
SMILES O=C(C1=CC(OC)=C(OCO2)C2=C1C3=C4OCOC4=C(OC)C=C3CO)OC
Molecular Weight (g/mol) 390.34
Formula Weight 390.34
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.