missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[4.1.0]heptane-7-carboxylic Acid, TRC

Product Code. 30354549
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354549 250 mg -
30368571 500 mg 500mg
30358805 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354549 Supplier Toronto Research Chemicals Supplier No. TRCC986323250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41894-76-2
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.17
Synonym NSC 156902
IUPAC Name bicyclo[4.1.0]heptane-7-carboxylic acid
SMILES OC(=O)C1C2CCCCC12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H12O2/c9-8(10)7-5-3-1-2-4-6(5)7/h5-7H,1-4H2,(H,9,10)
Formula Weight 140.0837
Chemical Name or Material Bicyclo[4.1.0]heptane-7-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.