missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[3.3.0]octane, Thermo Scientific™

Product Code. p-7238585 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 17525545

Brand: Thermo Scientific Acros 468522500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 694-72-4
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number 00060882
InChI Key AEBWATHAIVJLTA-UHFFFAOYSA-N
IUPAC Name octahydropentalene
SMILES C1CC2CCCC2C1

Specifications

Alpha Vector BICYCLOOCTANE
Color Colorless
Boiling Point 137°C
Flash Point 26°C
Infrared Spectrum Conforms
Packaging Glass Bottle
Quantity 250 mg
Physical Form Liquid
Chemical Name or Material Bicyclo[3.3.0]octane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.