missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[3.2.0]hept-2-en-6-one, 97%

Product Code. 11480647
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480647 100 g 100g
11460647 5 g 5g
11470647 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480647 Supplier Thermo Scientific Alfa Aesar Supplier No. B22472.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13173-09-6 | C7H8O | 108.14 g/mol

Bicyclo[3.2.0]hept-2-en-6-one is widely utilized to study enantioselective Baeyer-Villiger oxidation of (±)-cis-bicyclo(3.2.0)hept-2-en-6-one. It is used in the synthesis of a series of chalcone derivatives by reacting with arylaldehydes. It is also used to analyze the extremophile enzymes (monooxygenase and hydrolases) in microorganisms which are isolated from a deep-water petroleum reservoir and at high temperatures using fluorogenic assays and multibioreactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicyclo[3.2.0]hept-2-en-6-one is widely utilized to study enantioselective Baeyer-Villiger oxidation of (±)-cis-bicyclo(3.2.0)hept-2-en-6-one. It is used in the synthesis of a series of chalcone derivatives by reacting with arylaldehydes. It is also used to analyze the extremophile enzymes (monooxygenase and hydrolases) in microorganisms which are isolated from a deep-water petroleum reservoir and at high temperatures using fluorogenic assays and multibioreactions.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13173-09-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00066201
InChI Key LNLLHUHPGPKRBM-UHFFFAOYSA-N
Synonym bicyclo 3.2.0 hept-2-en-6-one, +/--cis-bicyclo 3.2.0 hept-2-en-6-one, cis-bicyclo 3.2.0 hept-2-en-6-one, 1s,5s-bicyclo 3.2.0 hept-2-en-6-one, +-1r,5s-cis-bicyclo 3.2.0 hept-2-en-6-one, bicyclo 3,2,0 hept-2-en-6-one, bicyclo 3,2,0 hept-2-ene-6-one, bicyclo 3.2.0 hept-3-en-7-one, z-bicyclo 3.2.0 hepta-2-ene-6-one
PubChem CID 297183
IUPAC Name bicyclo[3.2.0]hept-3-en-7-one
SMILES C1C=CC2C1C(=O)C2

Specifications

Quantity 100 g
UN Number UN1224
Sensitivity Air Sensitive
Formula Weight 108.14
Percent Purity 97%
Chemical Name or Material Bicyclo[3.2.0]hept-2-en-6-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.