missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 97%

Product Code. 11325279
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11325279 100 g 100g
11315279 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325279 Supplier Thermo Scientific Alfa Aesar Supplier No. A19632.22
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1719-83-1 | C12H8O6 | 248.19 g/mol

Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride is used to prepare the thermo-cured cycloaliphatic polyamic acid and photo-cured cycloaliphatic polyamic ester, which were then converted to cycloaliphatic polyimide film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride is used to prepare the thermo-cured cycloaliphatic polyamic acid and photo-cured cycloaliphatic polyamic ester, which were then converted to cycloaliphatic polyimide film.

Solubility
Soluble in water

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1719-83-1
Molecular Formula C12H8O6
Molecular Weight (g/mol) 248.19
MDL Number MFCD00082228
InChI Key XLOGCGOPKPCECW-UHFFFAOYSA-N
Synonym bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c:4,5-c' difuran-1,3,5,7 3ah,7ah-tetraone, bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic acid dianhydride, 4,8-etheno-1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, 3a,4,4a,7a,8,8a-hexahydro, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic dianhydride, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic acid dianhydride, 4,10-dioxatetracyclo 5.5.2.0<2,6>.0<8,12> tetradec-13-ene-3,5,9,11-tetraone, 4,10-dioxatetracyclo 5.5.2.0 2 ,?.0?, 1 2 tetradec-13-ene-3,5,9,11-tetrone, bicyclo 2.2.2 oct-7-ene-2,3:5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c
PubChem CID 102679
SMILES C1=CC2C3C(C1C4C2C(=O)OC4=O)C(=O)OC3=O

Specifications

Melting Point ∼360°C
Quantity 100 g
Beilstein 294213
Sensitivity Moisture sensitive
Solubility Information Soluble in water
Formula Weight 248.19
Percent Purity 97%
Chemical Name or Material Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.