missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 97%, ACROS Organics™

Product Code. 10367761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10367761 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10367761 Supplier Thermo Scientific Acros Supplier No. 176501000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1719-83-1
CAS Min % 96.0
CAS Max % 100.0
Color White to Beige
Elemental Analysis 56.00 to 60.10% (C)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C12H8O6
MDL Number MFCD00082228
Quantity 100 g
Synonym bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c:4,5-c' difuran-1,3,5,7 3ah,7ah-tetraone, bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic acid dianhydride, 4,8-etheno-1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, 3a,4,4a,7a,8,8a-hexahydro, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic dianhydride, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic acid dianhydride, 4,10-dioxatetracyclo 5.5.2.0<2,6>.0<8,12> tetradec-13-ene-3,5,9,11-tetraone, 4,10-dioxatetracyclo 5.5.2.0 2 ,?.0?, 1 2 tetradec-13-ene-3,5,9,11-tetrone, bicyclo 2.2.2 oct-7-ene-2,3:5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c
InChI Key XLOGCGOPKPCECW-UHFFFAOYSA-N
SMILES C1=CC2C3C(C1C4C2C(=O)OC4=O)C(=O)OC3=O
Molecular Weight (g/mol) 248.19
PubChem CID 102679
Formula Weight 248.19
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.