missing translation for 'onlineSavingsMsg'
Learn More

Bicuculline, MedChemExpress

Product Code. 16474787
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474787 50 mg 50mg
16429036 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16474787 Supplier Medchem Express Supplier No. HYN0219

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bicuculline ((+)-Bicuculline; d-Bicuculline), as a convulsant alkaloid, is a competitive neurotransmitter GABAA receptor antagonist (IC50=2 μM). Bicuculline also blocks Ca2+-activated potassium (SK) channels and subsequently blocks the slow afterhyperpolarization (slow AHP) .

Bicuculline ((+)-Bicuculline

Specifications

Chemical Name or Material Bicuculline
CAS 485-49-4
Color White
Purity Grade Notes Research
Molecular Formula C20H17NO6
Quantity 50 mg
Synonym (+)-Bicuculline d-Bicuculline
Solubility Information DMSO : 50 mg/mL (136.11 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1O[C@@H]([C@H]2N(C)CCC3=C2C=C(OCO4)C4=C3)C5=CC=C(OCO6)C6=C51
Molecular Weight (g/mol) 367.35
Formula Weight 367.35
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.